3-Bromo-5-(trifluoromethyl)aniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS40306G100EA 0 CHF
ACTIAS40306G100 ACTIAS40306G25
3-Bromo-5-(trifluoromethyl)aniline
3-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
MDL Number: MFCD00236205
CAS Number: 54962-75-3
EINECS: 259-412-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR