2-Bromo-5-(trifluoromethyl)aniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 6-Bromo-α,α,α-trifluoro-m-toluidine3-Amino-4-bromo-α,α,α-trifluorotoluene3-Amino-4-brom-α,α,α-trifluortoluol3-Amino-4-bromobenzotrifluoride

ACTIAS20584G100EA 0 CHF
ACTIAS20584G100 ACTIAS20584G25
2-Bromo-5-(trifluoromethyl)aniline
2-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 94…96 °C (12 mmHg)
Melting Pt: 47…49 °C
Density: 1.694 g/cm³ (25 °C)
MDL Number: MFCD00007637
CAS Number: 454-79-5
EINECS: 207-227-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR