2-Bromo-5-fluorotoluene

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 1-Bromo-4-fluoro-2-methylbenzene

ACTIAS21074G100EA 0 CHF
ACTIAS21074G100
2-Bromo-5-fluorotoluene
2-Bromo-5-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 177 °C (1013 hPa)
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00017921
CAS Number: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR