2-Bromo-5-chlorotoluene
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
ACTIAS32259G10EA
0
CHF
ACTIAS32259G10
2-Bromo-5-chlorotoluene
2-Bromo-5-chlorotoluene
Formula:
C₇H₆BrCl MW: 205.48 g/mol Boiling Pt: 98 °C (1013 hPa) Density: 1.55 g/cm³ (20 °C) |
MDL Number:
MFCD00000602 CAS Number: 14495-51-3 EINECS: 238-503-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...