α-Bromo-3-chlorotoulene

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: alpha-Bromo-3-chlorotoluene1-(Bromomethyl)-3-chlorobenzene3-Chlorobenzyl bromideα-Bromo-m-chlorotoulene

ACTIAS21082G100EA 0 CHF
ACTIAS21082G100
α-Bromo-3-chlorotoulene
α-Bromo-3-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 17.5 °C
Density: 1.56 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000597
CAS Number: 766-80-3
EINECS: 212-171-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR