1-Bromo-2-ethylbenzene
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS22497G100EA
0
CHF
ACTIAS22497G100
ACTIAS22497G25
1-Bromo-2-ethylbenzene
1-Bromo-2-ethylbenzene
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 199 °C (1013 hPa) Melting Pt: –67.3 °C Density: 1.3589 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000077 CAS Number: 1973-22-4 EINECS: 217-823-8 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...