4-Vinylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS16388G5EA 0 CHF
ACTIAS16388G5 ACTIAS16388G25
4-Vinylbenzoic acid
4-Vinylbenzoic acid
Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 115 °C
Density: 1,3 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
EINECS: 214-053-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR