3,4-Dimethoxyphenol
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS37545G10EA
0
CHF
ACTIAS37545G10
ACTIAS37545G5
3,4-Dimethoxyphenol
3,4-Dimethoxyphenol
Formel:
C₈H₁₀O₃ MW: 154,17 g/mol Siedepunkt: 120…122 °C (0,2 mmHg) Melting Pt: 83 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00008390 CAS-Nummer: 2033-89-8 EINECS: 217-995-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...