2-(2,4-Difluorophenyl)pyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS34586G5EA 0 CHF
ACTIAS34586G5 ACTIAS34586G10 ACTIAS34586G1
2-(2,4-Difluorophenyl)pyridine
2-(2,4-Difluorophenyl)pyridine
Formula: C₁₁H₇F₂N
MW: 191.18 g/mol
MDL Number: MFCD07787526
CAS Number: 391604-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR