1-Propylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50984G5EA 0 CHF
ACTIAS50984G1 ACTIAS50984G25 ACTIAS50984G5
1-Propylboronic acid
1-Propylboronic acid
Formula: C₃H₉BO₂
MW: 87.91 g/mol
MDL Number: MFCD01074564
CAS Number: 17745-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR