1-Octylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: n-Octylboronic acidOctylboronic acid1-n-Octaneboronic acid

ACTIAS51477G5EA 0 CHF
ACTIAS51477G5 ACTIAS51477G25 ACTIAS51477G1 ACTIAS51477G100
1-Octylboronic acid
1-Octylboronic acid
Formula: C₈H₁₉BO₂
MW: 158.05 g/mol
MDL Number: MFCD01074560
CAS Number: 28741-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR