5,6-Difluorooxindole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5,6-Difluoroindolin-2-one

ACTIAS36378G5EA 0 CHF
ACTIAS36378MG250 ACTIAS36378G5 ACTIAS36378G1
5,6-Difluorooxindole
5,6-Difluorooxindole
Formula: C₈H₅F₂NO
MW: 169.13 g/mol
MDL Number: MFCD01075216
CAS Number: 71294-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR