1-Benzyl-4-piperidone

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: N-Benzyl-4-piperidone1-Benzyl-4-piperidinone1-Benzylpiperidin-4-one

ACTIAS20094G100EA 0 CHF
ACTIAS20094G100 ACTIAS20094G500
1-Benzyl-4-piperidone
1-Benzyl-4-piperidone
Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
MDL Number: MFCD00006192
CAS Number: 3612-20-2
EINECS: 222-782-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR