(S)-(-)-4-Brom-α-methylbenzylalkohol

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS37160G1EA 0 CHF
ACTIAS37160MG250 ACTIAS37160G1
(S)-(-)-4-Brom-α-methylbenzylalkohol
(S)-(-)-4-Brom-α-methylbenzylalkohol
Formel: C₈H₉BrO
MW: 201,06 g/mol
MDL Number: MFCD06201860
CAS-Nummer: 100760-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR