5-Methoxytryptamin Hydrochlorid

ACTIAS21693G100EA 0 CHF
ACTIAS21693G100 ACTIAS21693G25
5-Methoxytryptamin Hydrochlorid
5-Methoxytryptamin Hydrochlorid
Formel: C₁₁H₁₅ClN₂O
MDL Number: MFCD00005662
CAS-Nummer: 66-83-1
EINECS: 200-637-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR