6-Fluorotryptamine hydrochloride

ACTIAS21190G1EA 0 CHF
ACTIAS21190G1 ACTIAS21190G5 ACTIAS21190MG250
6-Fluorotryptamine hydrochloride
6-Fluorotryptamine hydrochloride
Formula: C₁₀H₁₂ClFN₂
MW: 214.67 g/mol
MDL Number: MFCD00012687
CAS Number: 55206-24-1
EINECS: 259-533-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR