5-Chlorotryptamine hydrochloride

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 5-Chloro-3-(2-aminoethyl)indole Hydrochloride

ACTIAS32187G5EA 0 CHF
ACTIAS32187G5 ACTIAS32187G10 ACTIAS32187G1
5-Chlorotryptamine hydrochloride
5-Chlorotryptamine hydrochloride
Formula: C₁₀H₁₂Cl₂N₂
MW: 231.12 g/mol
MDL Number: MFCD00051990
CAS Number: 942-26-7
EINECS: 213-387-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR