3-Bromo-2-nitroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40316G5EA 0 CHF
ACTIAS40316G5
3-Bromo-2-nitroaniline
3-Bromo-2-nitroaniline
Formula: C₆H₅BrN₂O₂
MDL Number: MFCD08063248
CAS Number: 7138-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR