2-Amino-4-chlorbenzothiazol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS31442G5EA
0
CHF
ACTIAS31442G1
ACTIAS31442G5
2-Amino-4-chlorbenzothiazol
2-Amino-4-chlorobenzothiazole
Formel:
C₇H₅ClN₂S MW: 184,65 g/mol |
MDL Number:
MFCD00005791 CAS-Nummer: 19952-47-7 EINECS: 243-439-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...