2-Amino-5-brombenzothiazol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS30699G1EA
0
CHF
ACTIAS30699G1
ACTIAS30699G5
2-Amino-5-brombenzothiazol
2-Amino-5-bromobenzothiazole
Formel:
C₇H₅BrN₂S MW: 229,1 g/mol |
MDL Number:
MFCD00577806 CAS-Nummer: 20358-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...