Methylmalonic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Methylmalonic acid

ACTIAS22247G100EA 127 CHF
ACTIAS22247G500 ACTIAS22247G100
Methylmalonic acid
Methylmalonic acid
Formula: C₄H₆O₄
MW: 118.09 g/mol
MDL Number: MFCD00002656
CAS Number: 516-05-2
EINECS: 208-219-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR