1-Methyl-4-piperidone

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: N-Methyl-4-piperidone

ACTIAS20104G100EA 0 CHF
ACTIAS20104G100
1-Methyl-4-piperidone
1-Methyl-4-piperidone
Formula: C₆H₁₁NO
MW: 113.16 g/mol
MDL Number: MFCD00006191
CAS Number: 1445-73-4
EINECS: 215-895-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR