3-Nitrophenol
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
ACTIAS21081G100EA
0
CHF
ACTIAS21081G100
ACTIAS21081G25
3-Nitrophenol
3-Nitrophenol
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Boiling Pt: 271 °C (1013 hPa) Melting Pt: 94…95 °C Density: 1.485 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007240 CAS Number: 554-84-7 EINECS: 209-073-5 UN: 1663 ADR: 6.1,III Merck Index: 12,06716 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...