3,4-Dimethoxyaniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS20107G100EA 0 CHF
ACTIAS20107G100
3,4-Dimethoxyaniline
3,4-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 85…88 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008394
CAS Number: 6315-89-5
EINECS: 228-647-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR