3,4,5-Trifluorophenol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS33154G5EA 0 CHF
ACTIAS33154G10 ACTIAS33154G5
3,4,5-Trifluorophenol
3,4,5-Trifluorophenol
Formula: C₆H₃F₃O
MW: 148.08 g/mol
Density: 1.629 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042427
CAS Number: 99627-05-1
EINECS: 436-120-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR