3-Fluor-4-nitroanilin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS40351G5EA
0
CHF
ACTIAS40351G5
ACTIAS40351G10
3-Fluor-4-nitroanilin
3-Fluor-4-nitroanilin
Formel:
C₆H₅FN₂O₂ MW: 156,12 g/mol |
MDL Number:
MFCD03412239 CAS-Nummer: 2369-13-3 EINECS: 219-130-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...