3,5-Difluoroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS34298G100EA 0 CHF
ACTIAS34298G100 ACTIAS34298G10
3,5-Difluoroaniline
3,5-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 37…41 °C
Density: 1.295 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007763
CAS Number: 372-39-4
EINECS: 206-752-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR