2-Fluor-6-iodbenzoesäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS34859G5EA
0
CHF
ACTIAS34859G10
ACTIAS34859G100
ACTIAS34859G5
2-Fluor-6-iodbenzoesäure
2-Fluoro-6-iodobenzoic acid
Formel:
C₇H₄FIO₂ MW: 266,01 g/mol Siedepunkt: 312 °C (1013 hPa) Melting Pt: 123…126 °C |
MDL Number:
MFCD00042289 CAS-Nummer: 111771-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...