3,5-Difluorbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS31833G100EA 0 CHF
ACTIAS31833G100
3,5-Difluorbenzoesäure
3,5-Difluorobenzoic acid
Formel: C₇H₄F₂O₂
MW: 158,1 g/mol
Siedepunkt: 242 °C (1013 hPa)
Melting Pt: 122 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00010323
CAS-Nummer: 455-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR