2,3,4,5-Tetrafluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS32915G100EA 0 CHF
ACTIAS32915G100
2,3,4,5-Tetrafluorobenzoic acid
2,3,4,5-Tetrafluorobenzoic acid
Formula: C₇H₂F₄O₂
MW: 194.09 g/mol
Boiling Pt: 135…137 °C (23 mmHg)
Melting Pt: 92…92,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00009613
CAS Number: 1201-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR