2,4-Difluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS20249G100EA 121 CHF
ACTIAS20249G100
2,4-Difluorbenzonitril
2,4-Difluorbenzonitril
Formel: C₇H₃F₂N
MW: 139,1 g/mol
Siedepunkt: 80…83 °C (22,5 mmHg)
Melting Pt: 46…47 °C
MDL Number: MFCD00009826
CAS-Nummer: 3939-09-1
EINECS: 223-523-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR