5-Fluor-2-nitrobenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS40516G5EA
0
CHF
ACTIAS40516G5
5-Fluor-2-nitrobenzaldehyd
5-Fluor-2-nitrobenzaldehyd
Formel:
C₇H₄FNO₃ MW: 169,11 g/mol |
MDL Number:
MFCD00153175 CAS-Nummer: 395-81-3 EINECS: 206-903-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...