2,3-Difluorobenzaldehyde
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
ACTIAS31488G5EA
0
CHF
ACTIAS31488G5
ACTIAS31488G10
2,3-Difluorobenzaldehyde
2,3-Difluorobenzaldehyde
Formula:
C₇H₄F₂O MW: 142.1 g/mol Boiling Pt: 64…65 °C (17 mmHg) Density: 1.3 g/cm³ (20 °C) |
MDL Number:
MFCD00010292 CAS Number: 2646-91-5 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...