2-Chloro-5-nitroanisole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31124G100EA 0 CHF
ACTIAS31124G100
2-Chloro-5-nitroanisole
2-Chloro-5-nitroanisole
Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
MDL Number: MFCD00079739
CAS Number: 1009-36-5
EINECS: 213-768-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR