2,6-Dichloroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

ACTIAS32769G100EA 0 CHF
ACTIAS32769G100
2,6-Dichloroaniline
2,6-Dichloroaniline
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 35…38 °C
Density: 1.526 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007675
CAS Number: 608-31-1
EINECS: 210-160-5
UN: 3442
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR