2-Chloro-1-fluoro-4-nitrobenzene

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Chloro-2-fluoro-5-nitrobenzene3-Chloro-4-fluoronitrobenzene

ACTIAS20137G100EA 0 CHF
ACTIAS20137G500 ACTIAS20137G100
2-Chloro-1-fluoro-4-nitrobenzene
2-Chloro-1-fluoro-4-nitrobenzene
Formula: C₆H₃ClFNO₂
MW: 175.55 g/mol
Boiling Pt: 227…228 °C (1013 hPa)
Melting Pt: 41.5 °C
MDL Number: MFCD00007206
CAS Number: 350-30-1
EINECS: 206-499-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR