2-Chloro-4-fluoroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS33667G100EA 0 CHF
ACTIAS33667G10 ACTIAS33667G100
2-Chloro-4-fluoroaniline
2-Chloro-4-fluoroaniline
Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 192 °C (1013 hPa)
Density: 1.21 g/cm³ (20 °C)
MDL Number: MFCD00042530
CAS Number: 2106-02-7
EINECS: 218-282-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR