2-Chloro-6-fluorobenzoic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
ACTIAS31547G100EA
0
CHF
ACTIAS31547G100
2-Chloro-6-fluorobenzoic acid
2-Chloro-6-fluorobenzoic acid
Formula:
C₇H₄ClFO₂ MW: 174.56 g/mol Boiling Pt: 272 °C (1013 hPa) Melting Pt: 157…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002417 CAS Number: 434-75-3 EINECS: 207-105-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...