2-Chlor-4,5-difluorbenzoesäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Gefahr
|
ACTIAS31135G5EA
0
CHF
ACTIAS31135G5
ACTIAS31135G10
2-Chlor-4,5-difluorbenzoesäure
2-Chloro-4,5-difluorobenzoic acid
Formel:
C₇H₃ClF₂O₂ MW: 192,55 g/mol Siedepunkt: 258 °C (1013 hPa) Melting Pt: 103…106 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00077479 CAS-Nummer: 110877-64-0 EINECS: 405-380-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...