4-Chloro-3-fluorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS40425G5EA 0 CHF
ACTIAS40425G5
4-Chloro-3-fluorobenzaldehyde
4-Chloro-3-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
MDL Number: MFCD00143288
CAS Number: 5527-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR