5-Amino-2-chlorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS32507G100EA 0 CHF
ACTIAS32507G100
5-Amino-2-chlorobenzoic acid
5-Amino-2-chlorobenzoic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1,519 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007845
CAS Number: 89-54-3
EINECS: 201-916-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR