6-Chloranthranilsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS20162G25EA 0 CHF
ACTIAS20162G25 ACTIAS20162G100
6-Chloranthranilsäure
6-Chloranthranilsäure
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Siedepunkt: 330 °C (1013 hPa)
Melting Pt: 153…154 °C
MDL Number: MFCD00051530
CAS-Nummer: 2148-56-3
EINECS: 218-416-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR