5-Chloroanthranilic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
Synonyms:
2-Amino-5-chlorobenzoic acid
ACTIAS20116G100EA
0
CHF
ACTIAS20116G100
5-Chloroanthranilic acid
5-Chloroanthranilic acid
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007838 CAS Number: 635-21-2 EINECS: 211-230-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...