3,5-Dichlorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS32992G100EA 0 CHF
ACTIAS32992G100
3,5-Dichlorobenzoic acid
3,5-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00002494
CAS Number: 51-36-5
EINECS: 200-092-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR