4-Chloro-3-nitrobenzonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS22096G25EA 84 CHF
ACTIAS22096G25
4-Chloro-3-nitrobenzonitrile
4-Chloro-3-nitrobenzonitrile
Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00016987
CAS Number: 939-80-0
EINECS: 213-364-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR