3-Chlorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS20239G100EA 0 CHF
ACTIAS20239G100 ACTIAS20239G25
3-Chlorobenzaldehyde
3-Chlorobenzaldehyde
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 17…18 °C
Density: 1,241 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR