3,5-Dichlorbenzaldehyd
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS20101G25EA
0
CHF
ACTIAS20101G25
3,5-Dichlorbenzaldehyd
3,5-Dichlorobenzaldehyde
Formel:
C₇H₄Cl₂O MW: 175,01 g/mol Siedepunkt: 215…222 °C (7 mmHg) Melting Pt: 63 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003352 CAS-Nummer: 10203-08-4 EINECS: 233-499-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...