3,4-Dichlorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

ACTIAS20161G100EA 0 CHF
ACTIAS20161G100
3,4-Dichlorobenzaldehyde
3,4-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003351
CAS Number: 6287-38-3
EINECS: 228-520-5
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR