2,3-Dihydroxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS20844G25EA 0 CHF
ACTIAS20844G25 ACTIAS20844G100
2,3-Dihydroxybenzoic acid
2,3-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 203…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00002446
CAS Number: 303-38-8
EINECS: 206-139-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR