Syringic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Hydroxy-3,5-dimethoxybenzoic acid

ACTIAS21908G100EA 0 CHF
ACTIAS21908G25 ACTIAS21908G100
Syringic acid
Syringic acid
Formula: C₉H₁₀O₅
MW: 198.18 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00002552
CAS Number: 530-57-4
EINECS: 208-486-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR